accessibility menu, dialog, popup

UserName

CdOHOH

CAS RN 21041-95-2

IUPAC Name: cadmium hydroxide
Synonyms: cadmium hydroxide
Molecular Weight (g/mol): 148.44
Molecular Formula: CdH4O2
InChi Key: XIYPBSADJSJKMF-UHFFFAOYSA-N
SMILES: O.O.[Cd]
Grade 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)