accessibility menu, dialog, popup

UserName

CAS RN 21020-27-9

CH3H3CH3C

CAS RN 21020-27-9

IUPAC Name: 4-methylpent-2-yne
Synonyms: 4-methylpent-2-yne
Molecular Weight (g/mol): 82.15
Molecular Formula: C6H10
InChi Key: SLMFWJQZLPEDDU-UHFFFAOYSA-N
SMILES: CC#CC(C)C
Boiling Point 
  • (5)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)