accessibility menu, dialog, popup

UserName

H3COOCH3Cl

CAS RN 2100-42-7

IUPAC Name: 2-chloro-1,4-dimethoxybenzene
Synonyms: 1,4-dimethoxy-2-chlorobenzene
Molecular Weight (g/mol): 172.61
Molecular Formula: C8H9ClO2
InChi Key: QMXZSRVFIWACJH-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results