accessibility menu, dialog, popup

UserName

CAS RN 20996-62-7

IUPAC Name: 1-(ethylsulfanyl)propan-2-one
Synonyms: 1-(ethylsulfanyl)propan-2-one
Molecular Weight (g/mol): 118.19
Molecular Formula: C5H10OS
InChi Key: ZKDXKBIOEVBFGV-UHFFFAOYSA-N
SMILES: CCSCC(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results