accessibility menu, dialog, popup

UserName

CAS RN 209733-38-0

IUPAC Name: molybdenum pentakis(propan-2-olate)
Synonyms: molybdenum pentakis(propan-2-olate)
Molecular Weight (g/mol): 391.39
Molecular Formula: C15H35MoO5
InChi Key: WMGOKFNZAFRBMJ-UHFFFAOYSA-N
SMILES: [Mo].CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-]
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)