accessibility menu, dialog, popup

UserName

NHNNSHCl

CAS RN 209733-13-1

IUPAC Name: 1-(1,3-thiazol-2-yl)piperazine hydrochloride
Synonyms: piperazine, 1-(2-thiazolyl)- hydrochloride
Molecular Weight (g/mol): 205.7
Molecular Formula: C7H12ClN3S
InChi Key: BIHQYKGSYPXHRN-UHFFFAOYSA-N
SMILES: Cl.C1CN(CCN1)C1=NC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results