accessibility menu, dialog, popup

UserName

CH3SON

CAS RN 20949-84-2

IUPAC Name: 2-methyl-1,3-thiazole-4-carbaldehyde
Synonyms: 2-methyl-1,3-thiazole-4-carbaldehyde
Molecular Weight (g/mol): 127.16
Molecular Formula: C5H5NOS
InChi Key: AEHWVNPVEUVPMT-UHFFFAOYSA-N
SMILES: CC1=NC(C=O)=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results