accessibility menu, dialog, popup

UserName

CH3SSOHO

CAS RN 20873-58-9

IUPAC Name: 5-(methylsulfanyl)thiophene-2-carboxylic acid
Synonyms: 5-(methylsulfanyl)thiophene-2-carboxylic acid
Molecular Weight (g/mol): 174.23
Molecular Formula: C6H6O2S2
InChi Key: FWFWUSLBIIIIEN-UHFFFAOYSA-N
SMILES: CSC1=CC=C(S1)C(O)=O
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results