accessibility menu, dialog, popup

UserName

CAS RN 2080-75-3

CH3OHNOHN

CAS RN 2080-75-3

IUPAC Name: 5-methoxy-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: 5-methoxy-1,3-dihydro-1,3-benzodiazol-2-one
Molecular Weight (g/mol): 164.16
Molecular Formula: C8H8N2O2
InChi Key: AUPLVAKFTYFHTA-UHFFFAOYSA-N
SMILES: COC1=CC=C2NC(=O)NC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results