accessibility menu, dialog, popup

UserName

FFFFO

CAS RN 207853-70-1

IUPAC Name: [2-fluoro-3-(trifluoromethyl)phenyl](phenyl)methanone
Synonyms: [2-fluoro-3-(trifluoromethyl)phenyl](phenyl)methanone
Molecular Weight (g/mol): 268.21
Molecular Formula: C14H8F4O
InChi Key: QNEOCRQLKSEYEM-UHFFFAOYSA-N
SMILES: FC1=C(C=CC=C1C(F)(F)F)C(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results