accessibility menu, dialog, popup

UserName

CAS RN 20776-50-5

IUPAC Name: 2-amino-4-bromobenzoic acid
Synonyms: 2-amino-4-bromobenzoic acid
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: BNNICQAVXPXQAH-UHFFFAOYSA-N
SMILES: NC1=CC(Br)=CC=C1C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results