accessibility menu, dialog, popup

UserName

CAS RN 207596-32-5

NOOONOOOPd

CAS RN 207596-32-5

IUPAC Name: palladium(2+) hydrate dinitrate
Synonyms: palladium(2+) hydrate dinitrate
Molecular Weight (g/mol): 248.44
Molecular Formula: H2N2O7Pd
InChi Key: DTZRLFJKQHIVQA-UHFFFAOYSA-N
SMILES: O.[Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Physical Form 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)