accessibility menu, dialog, popup

UserName

H3CSOOOH2ONa

CAS RN 207596-29-0

IUPAC Name: sodium hydrate octane-1-sulfonate
Synonyms: sodium 1-octanesulfonate hydrate
Molecular Weight (g/mol): 234.29
Molecular Formula: C8H19NaO4S
InChi Key: MBURIAHQXJQKRE-UHFFFAOYSA-M
SMILES: O.[Na+].CCCCCCCCS([O-])(=O)=O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)