accessibility menu, dialog, popup

UserName

H3CSOOOH2ONa

CAS RN 207300-90-1

IUPAC Name: sodium heptane-1-sulfonate hydrate
Synonyms: sodium heptane-1-sulfonate hydrate
Molecular Weight (g/mol): 220.26
Molecular Formula: C7H17NaO4S
InChi Key: XWZCREJRXRKIRQ-UHFFFAOYSA-M
SMILES: O.[Na+].CCCCCCCS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Physical Form 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (1)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)