CAS RN 2070009-72-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MMAE-d8, MedChemExpress
MedChemExpress MMAE-d8 (Monomethyl auristatin E-d8) is a deuterated labeled MMAE, a potent mitotic inhibitor and a tubulin inhibitor.