accessibility menu, dialog, popup

UserName

H3CH3CCH3OH

CAS RN 20698-30-0

IUPAC Name: 4-(2-methylbutan-2-yl)cyclohexan-1-ol
Synonyms: tert-amylcyclohexanol
Molecular Weight (g/mol): 170.3
Molecular Formula: C11H22O
InChi Key: VDQZABQVXYELSI-UHFFFAOYSA-N
SMILES: CCC(C)(C)C1CCC(O)CC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results