accessibility menu, dialog, popup

UserName

CAS RN 2068-24-8

HOOSSOHO

CAS RN 2068-24-8

IUPAC Name: 2-({[(carboxymethyl)sulfanyl]methyl}sulfanyl)acetic acid
Synonyms: ({[(carboxymethyl)sulfanyl]methyl}sulfanyl)acetic acid
Molecular Weight (g/mol): 196.24
Molecular Formula: C5H8O4S2
InChi Key: CZEVAMMETWVBOD-UHFFFAOYSA-N
SMILES: OC(=O)CSCSCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results