accessibility menu, dialog, popup

UserName

FNCSFF

CAS RN 206761-91-3

IUPAC Name: 1,3,5-trifluoro-2-isothiocyanatobenzene
Synonyms: 1,3,5-trifluoro-2-isothiocyanatobenzene
Molecular Weight (g/mol): 189.16
Molecular Formula: C7H2F3NS
InChi Key: JPISVBDECFJHDB-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(N=C=S)C(F)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results