accessibility menu, dialog, popup

UserName

CAS RN 20662-53-7

OHNNHN

CAS RN 20662-53-7

IUPAC Name: 4-(2-oxo-2,3-dihydro-1H-1,3-benzodiazol-1-yl)piperidin-1-ium
Synonyms: 4-(2-oxo-3H-1,3-benzodiazol-1-yl)piperidin-1-ium
Molecular Weight (g/mol): 218.28
Molecular Formula: C12H16N3O
InChi Key: BYNBAMHAURJNTR-UHFFFAOYSA-O
SMILES: O=C1NC2=CC=CC=C2N1C1CC[NH2+]CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results