CAS RN 2065250-25-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
DAAO inhibitor-1, MedChemExpress
MedChemExpress DAAO inhibitor-1 is a potent D-amino acid oxidase (DAAO) inhibitor with an IC50 of 0.12 μM.