accessibility menu, dialog, popup

UserName

CAS RN 20634-12-2

NNNNPtONOOONOO

CAS RN 20634-12-2

IUPAC Name: platinum(2+) tetraamine dinitrate
Synonyms: platinum(2+) tetraamine dinitrate
Molecular Weight (g/mol): 387.22
Molecular Formula: H12N6O6Pt
InChi Key: RBAKORNXYLGSJB-UHFFFAOYSA-N
SMILES: N.N.N.N.[Pt++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Physical Form 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)