accessibility menu, dialog, popup

UserName

CH3OHNSOOH3C

CAS RN 20628-27-7

IUPAC Name: N-(2-methanesulfonylphenyl)acetamide
Synonyms: N-(2-methanesulfonylphenyl)acetamide
Molecular Weight (g/mol): 213.25
Molecular Formula: C9H11NO3S
InChi Key: NGWCPCSZLLJKPD-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC=C1S(C)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results