accessibility menu, dialog, popup

UserName

CH3OONClBr

CAS RN 206257-39-8

IUPAC Name: ethyl 6-bromo-4-chloroquinoline-3-carboxylate
Synonyms: ethyl 6-bromo-4-chloroquinoline-3-carboxylate
Molecular Weight (g/mol): 314.56
Molecular Formula: C12H9BrClNO2
InChi Key: YGMLKBFPHVLHGT-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C2C=CC(Br)=CC2=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results