accessibility menu, dialog, popup

UserName

CAS RN 205445-52-9

H3CH3CCH3OONH(S)FFOHO

CAS RN 205445-52-9

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(3,5-difluorophenyl)propanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-(3,5-difluorophenyl)propanoic acid
Molecular Weight (g/mol): 301.29
Molecular Formula: C14H17F2NO4
InChi Key: CZBNUDVCRKSYDG-NSHDSACASA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC(F)=CC(F)=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results