accessibility menu, dialog, popup

UserName

CAS RN 20532-33-6

SCl

CAS RN 20532-33-6

IUPAC Name: 5-chloro-1-benzothiophene
Synonyms: 5-chloro-1-benzothiophene
Molecular Weight (g/mol): 168.64
Molecular Formula: C8H5ClS
InChi Key: SNYURIHMNFPQFL-UHFFFAOYSA-N
SMILES: ClC1=CC=C2SC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results