accessibility menu, dialog, popup

UserName

CAS RN 20532-28-9

NH2S

CAS RN 20532-28-9

IUPAC Name: 1-benzothiophen-5-amine
Synonyms: 1-benzothiophen-5-amine
Molecular Weight (g/mol): 149.21
Molecular Formula: C8H7NS
InChi Key: ZUPYTANKWDPRDP-UHFFFAOYSA-N
SMILES: NC1=CC=C2SC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results