CAS RN 2051-98-1
CAS RN 2051-98-1
IUPAC Name:
5-bromo-1,2-dihydroacenaphthylene
Synonyms:
5-bromo-1,2-dihydroacenaphthylene
Molecular Weight (g/mol):
233.11
Molecular Formula:
C12H9Br
InChi Key:
QALKJGMGKYKMKE-UHFFFAOYSA-N
SMILES:
BrC1=C2C=CC=C3CCC(C=C1)=C23
1
–
1
of
1
results
1
5-Bromoacenaphthene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.