accessibility menu, dialog, popup

UserName

CAS RN 2051-90-3

ClCl

CAS RN 2051-90-3

IUPAC Name: [dichloro(phenyl)methyl]benzene
Synonyms: diphenyldichloromethane
Molecular Weight (g/mol): 237.12
Molecular Formula: C13H10Cl2
InChi Key: OPTDDWCXQQYKGU-UHFFFAOYSA-N
SMILES: ClC(Cl)(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)