accessibility menu, dialog, popup

UserName

CAS RN 2051-49-2

H3COOOCH3

CAS RN 2051-49-2

IUPAC Name: hexanoyl hexanoate
Synonyms: hexanoic acid, anhydride
Molecular Weight (g/mol): 214.31
Molecular Formula: C12H22O3
InChi Key: PKHMTIRCAFTBDS-UHFFFAOYSA-N
SMILES: CCCCCC(=O)OC(=O)CCCCC
Boiling Point 
  • (5)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)