accessibility menu, dialog, popup

UserName

HN

CAS RN 2047-89-4

IUPAC Name: 5H,6H,7H,8H,9H,10H-cyclohepta[b]indole
Synonyms: 5H,6H,7H,8H,9H,10H-cyclohepta[b]indole
Molecular Weight (g/mol): 185.27
Molecular Formula: C13H15N
InChi Key: XZUJMYLNFZHNLP-UHFFFAOYSA-N
SMILES: C1CCC2=C(CC1)C1=CC=CC=C1N2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results