accessibility menu, dialog, popup

UserName

CAS RN 20445-33-4

IUPAC Name: (2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
Synonyms: (2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
Molecular Weight (g/mol): 252.62
Molecular Formula: C10H8ClF3O2
InChi Key: PAORVUMOXXAMPL-SECBINFHSA-N
SMILES: CO[C@@](C(Cl)=O)(C1=CC=CC=C1)C(F)(F)F
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)