accessibility menu, dialog, popup

UserName

H3COHOOFFF

CAS RN 20445-31-2

IUPAC Name: 3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid
Synonyms: mosher's acid
Molecular Weight (g/mol): 234.17
Molecular Formula: C10H9F3O3
InChi Key: JJYKJUXBWFATTE-UHFFFAOYNA-N
SMILES: COC(C(O)=O)(C1=CC=CC=C1)C(F)(F)F
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)