accessibility menu, dialog, popup

UserName

CAS RN 2040-01-9

H3COCH3CH3CH3H3CH3C

CAS RN 2040-01-9

IUPAC Name: 1-(2,3,4,5,6-pentamethylphenyl)ethan-1-one
Synonyms: 1-(2,3,4,5,6-pentamethylphenyl)ethanone
Molecular Weight (g/mol): 190.29
Molecular Formula: C13H18O
InChi Key: CTTYWXDVWGKHKJ-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(C)C(C)=C(C)C(C)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results