accessibility menu, dialog, popup

UserName

CAS RN 20388-87-8

HClCH3NH2

CAS RN 20388-87-8

IUPAC Name: hydrogen 2-phenylpropan-1-amine chloride
Synonyms: hydrogen Β-methylphenethylamine chloride
Molecular Weight (g/mol): 171.67
Molecular Formula: C9H14ClN
InChi Key: HBVYOCJBEXSCQE-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(CN)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)