accessibility menu, dialog, popup

UserName

CAS RN 20375-65-9

ClNN

CAS RN 20375-65-9

IUPAC Name: 3-chloro-6-phenylpyridazine
Synonyms: 3-chloro-6-phenylpyridazine
Molecular Weight (g/mol): 190.63
Molecular Formula: C10H7ClN2
InChi Key: BUBRFWDEAVIFMV-UHFFFAOYSA-N
SMILES: ClC1=CC=C(N=N1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results