accessibility menu, dialog, popup

UserName

SSn

CAS RN 20332-10-9

IUPAC Name: diphenylstannanethione
Synonyms: diphenylstannanethione
Molecular Weight (g/mol): 304.98
Molecular Formula: C12H10SSn
InChi Key: CYXRBVIJBDYNQU-UHFFFAOYSA-N
SMILES: S=[Sn](C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results