accessibility menu, dialog, popup

UserName

CAS RN 2032-65-7

H3CNHOOCH3SCH3H3C

CAS RN 2032-65-7

IUPAC Name: 3,5-dimethyl-4-(methylsulfanyl)phenyl N-methylcarbamate
Synonyms: grandslam mesurol methiocarb certan caswell no. 578B lizetan epa pesticide chemical code 100501 mesurol phenol draza mercaptodimethur
Molecular Weight (g/mol): 225.31
Molecular Formula: C11H15NO2S
InChi Key: YFBPRJGDJKVWAH-UHFFFAOYSA-N
SMILES: CNC(=O)OC1=CC(C)=C(SC)C(C)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (1)
Solvent or Matrix 
  • (1)
  • (2)
Concentration 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Element 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)