accessibility menu, dialog, popup

UserName

H3COSiCH3OCH3

CAS RN 2031-62-1

IUPAC Name: diethoxy(methyl)silyl
Synonyms: diethoxy(methyl)silyl
Molecular Weight (g/mol): 133.24
Molecular Formula: C5H13O2Si
InChi Key: GAURFLBIDLSLQU-UHFFFAOYSA-N
SMILES: CCO[Si](C)OCC
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results