accessibility menu, dialog, popup

UserName

CAS RN 20277-69-4

IUPAC Name: sodium methanesulfinate
Synonyms: sodium methanesulfinate
Molecular Weight (g/mol): 102.08
Molecular Formula: CH3NaO2S
InChi Key: LYPGDCWPTHTUDO-UHFFFAOYSA-M
SMILES: [Na+].CS([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)