CAS RN 20211-76-1
CAS RN 20211-76-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Lanthanum chloride, hydrated, 64.5-70% LaCl3
CAS: 20211-76-1 Molecular Formula: Cl3La Molecular Weight (g/mol): 245.26 MDL Number: MFCD00149753 InChI Key: ICAKDTKJOYSXGC-UHFFFAOYSA-K IUPAC Name: lanthanum(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[La+3]
3
Lanthanum(III) chloride hydrate, 99.9% (REO)
CAS: 20211-76-1 Molecular Formula: Cl3La Molecular Weight (g/mol): 245.26 MDL Number: MFCD00149753 InChI Key: ICAKDTKJOYSXGC-UHFFFAOYSA-K IUPAC Name: lanthanum(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[La+3]
4
Lanthanum(III) chloride hydrate, Honeywell™
CAS: 20211-76-1 Molecular Formula: Cl3La Molecular Weight (g/mol): 245.26 MDL Number: MFCD00149753 InChI Key: ICAKDTKJOYSXGC-UHFFFAOYSA-K IUPAC Name: lanthanum(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[La+3]