accessibility menu, dialog, popup

UserName

H3COOOCH3CH3

CAS RN 20201-24-5

IUPAC Name: ethyl 3-methyl-2-oxobutanoate
Synonyms: ethyl 3-methyl-2-oxobutanoate
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: CKTYYUQUWFEUCO-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)C(C)C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results