CAS RN 201943-63-7
CAS RN 201943-63-7
IUPAC Name:
2-[1-amino-1-carboxy-2-(9H-xanthen-9-yl)ethyl]cyclopropane-1-carboxylic acid
Synonyms:
2-[1-amino-1-carboxy-2-(9H-xanthen-9-yl)ethyl]cyclopropane-1-carboxylic acid
Molecular Weight (g/mol):
353.37
Molecular Formula:
C20H19NO5
InChi Key:
VLZBRVJVCCNPRJ-UHFFFAOYNA-N
SMILES:
NC(CC1C2=CC=CC=C2OC2=CC=CC=C12)(C1CC1C(O)=O)C(O)=O
1
–
2
of
2
results
1
Tocris Bioscience™ LY 341495
CAS: 201943-63-7 Molecular Formula: C20H19NO5
2
LY341495, MedChemExpress
MedChemExpress LY341495 is a metabotropic glutamate receptor (mGluR) antagonist with IC50s of 21 nM, 14 nM, 7.8 μM, 8.2 μM, 170 nM, 990 nM, 22 μM for mGlu2, mGlu3, mGlu1a, mGlu5a, mGlu8, mGlu7, and mGlu4 receptors, respectively.