accessibility menu, dialog, popup

UserName

CAS RN 201802-67-7

BNHOOH

CAS RN 201802-67-7

IUPAC Name: [4-(diphenylamino)phenyl]boronic acid
Synonyms: 4-(diphenylamino)phenylboronic acid
Molecular Weight (g/mol): 289.14
Molecular Formula: C18H16BNO2
InChi Key: TWWQCBRELPOMER-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results