accessibility menu, dialog, popup

UserName

CAS RN 2018-61-3

IUPAC Name: (2S)-2-acetamido-3-phenylpropanoic acid
Synonyms: acetyl-L-phenylalanine N-acetyl-l-phenalanine
Molecular Weight (g/mol): 207.23
Molecular Formula: C11H13NO3
InChi Key: CBQJSKKFNMDLON-JTQLQIEISA-N
SMILES: CC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results