CAS RN 201341-05-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tenofovir Disoproxil, MedChemExpress
MedChemExpress Tenofovir Disoproxil (Bis(POC)-PMPA) is a nucleotide reverse transcriptase inhibitor to treat HIV and chronic Hepatitis B.