Learn More
Tenofovir Disoproxil, MedChemExpress
Product Code. 16486326
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486326 100 mg 100mg
16481706 500 mg 500mg
16461106 200 mg 200mg
16452007 50 mg 50mg
16445858 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16486326 Supplier Medchem Express Supplier No. HY13782A
MedChemExpress Tenofovir Disoproxil (Bis(POC)-PMPA) is a nucleotide reverse transcriptase inhibitor to treat HIV and chronic Hepatitis B.
Tenofovir Disoproxil (Bis(POC)-PMPA) is a nucleotide reverse transcriptase inhibitor to treat HIV and chronic Hepatitis B.
Specifications
Chemical Name or Material Tenofovir Disoproxil
Physical Form Solid
CAS 201341-05-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H30N5O10P
For Use With (Application) Neuroscience-Neuromodulation
Synonym Bis(POC)-PMPA GS 4331
Solubility Information DMSO : ≥ 38 mg/mL (73.16 mM)
SMILES O=C(OC(C)C)OCOP(OCOC(OC(C)C)=O)(CO[C@H](C)CN1C=NC2=C(N)N=CN=C12)=O
Molecular Weight (g/mol) 519.44
Formula Weight 519.44
Percent Purity 99.84%
Grade Research