accessibility menu, dialog, popup

UserName

CH3OOH

CAS RN 201058-08-4

IUPAC Name: {4-[(4-methylphenyl)methoxy]phenyl}methanol
Synonyms: {4-[(4-methylphenyl)methoxy]phenyl}methanol
Molecular Weight (g/mol): 228.29
Molecular Formula: C15H16O2
InChi Key: NERFNHBZJXXFGY-UHFFFAOYSA-N
SMILES: CC1=CC=C(COC2=CC=C(CO)C=C2)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)