accessibility menu, dialog, popup

UserName

CAS RN 19968-85-5

ClNH2HO

CAS RN 19968-85-5

IUPAC Name: 1-(aminomethyl)cyclohexan-1-ol hydrochloridyl
Synonyms: 1-(aminomethyl)cyclohexan-1-ol hydrochloridyl
Molecular Weight (g/mol): 164.65
Molecular Formula: C7H15ClNO
InChi Key: FPVJGAVWTOENOY-UHFFFAOYSA-N
SMILES: [Cl].NCC1(O)CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results