accessibility menu, dialog, popup

UserName

CAS RN 19968-17-3

NHNBrFFF

CAS RN 19968-17-3

IUPAC Name: 4-bromo-5-(trifluoromethyl)-1H-pyrazole
Synonyms: 4-bromo-3-(trifluoromethyl)-2H-pyrazole
Molecular Weight (g/mol): 214.97
Molecular Formula: C4H2BrF3N2
InChi Key: JTHNMRUVJDWVMJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(Br)C=NN1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)