accessibility menu, dialog, popup

UserName

H2NSClN

CAS RN 19952-47-7

IUPAC Name: 4-chloro-1,3-benzothiazol-2-amine
Synonyms: 2-benzothiazolamine, 4-chloro-
Molecular Weight (g/mol): 184.64
Molecular Formula: C7H5ClN2S
InChi Key: OEQQFQXMCPMEIH-UHFFFAOYSA-N
SMILES: NC1=NC2=C(Cl)C=CC=C2S1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)